CHIS5-06095

5-06096 5-06095
76230-326EA 301.34 USD
76230-326 76230-324
CHIS5-06095
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR