CHIS5-06047

5-06048 5-06047
76230-238EA 589.57 USD
76230-238 76230-236
CHIS5-06047
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR