CHIS5-06043

5-06044 5-06043
76233-596EA 163.77 USD
76233-596 76233-598
CHIS5-06043
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR