CHIS5-06035

5-06036 5-06035
76230-220EA 163.77 USD
76230-220 76230-222
CHIS5-06035
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR