CHIS5-06021

5-06022 5-06021
76230-194EA 206.35 USD
76230-194 76230-192
CHIS5-06021
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR