CHIS5-06017

5-06018 5-06017
76230-186EA 245.66 USD
76230-186 76230-184
CHIS5-06017
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR