CHIS5-05971

5-05971 5-05972
76230-038EA 517.51 USD
76230-038 76230-036
CHIS5-05971
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR