CHIS5-05967

5-05968 5-05967
76230-028EA 180.15 USD
76230-028 76230-030
CHIS5-05967
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR