CHIS5-05913

5-05913 5-05914
76226-306EA 294.79 USD
76226-306 76226-304
CHIS5-05913
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR