CHIS5-05905

5-05905 5-05906
76226-288EA 86.8 USD
76226-288 76226-290
CHIS5-05905
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR