CHIS5-05899

5-05899 5-05900
76226-276EA 131.02 USD
76226-276 76226-278
CHIS5-05899
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR