CHIS5-05897

5-05897 5-05898
76226-274EA 262.03 USD
76226-274 76226-272
CHIS5-05897
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR