CHIS5-05865

5-05865 5-05866
76226-232EA 131.02 USD
76226-232 76226-234
CHIS5-05865
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR