CHIS5-05863

5-05863 5-05864
76226-230EA 343.92 USD
76226-230 76226-228
CHIS5-05863
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR