CHIS5-05861

5-05861 5-05862
76226-224EA 217.82 USD
76226-224 76226-226
CHIS5-05861
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR