CHIS5-05851

5-05851 5-05852
76229-964EA 223.55 USD
76229-964 76229-966
CHIS5-05851
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR