CHIS5-05829

5-05829 5-05830
76229-114EA 327.54 USD
76229-114 76229-112
CHIS5-05829
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR