CHIS5-05821

5-05821 5-05822
76226-642EA 900.74 USD
76226-642 76226-640
CHIS5-05821
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR