CHIS5-05815

5-05815 5-05816
76229-102EA 2407.42 USD
76229-102 76229-100
CHIS5-05815
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR