CHIS5-05811

5-05812 5-05811
76229-092EA 81.89 USD
76229-092 76229-094
CHIS5-05811
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR