CHIS5-05799

5-05799 5-05800
76229-070EA 343.92 USD
76229-070 76229-068
CHIS5-05799
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR