CHIS5-05769

5-05769 5-05770
76226-162EA 818.85 USD
76226-162 76226-160
CHIS5-05769
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR