CHIS5-05755

5-05755 5-05756
76229-044EA 394.69 USD
76229-044 76229-046
CHIS5-05755
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR