CHIS5-05745

5-05746 5-05745
76229-026EA 212.9 USD
76229-026 76229-024
CHIS5-05745
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR