CHIS5-05739

5-05739 5-05740
76229-014EA 188.34 USD
76229-014 76229-012
CHIS5-05739
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR