CHIS5-05733

5-05734 5-05733
76229-000EA 50.77 USD
76229-000 76229-002
CHIS5-05733
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR