CHIS5-05717

5-05718 5-05717
76228-986EA 859.8 USD
76228-986 76228-984
CHIS5-05717
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR