CHIS5-05685

5-05685 5-05686
76226-136EA 196.52 USD
76226-136 76226-138
CHIS5-05685
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR