CHIS5-05655

5-05656 5-05655
76228-956EA 49.13 USD
76228-956 76226-102
CHIS5-05655
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR