CHIS5-05639

5-05639 5-05640
76226-494EA 1228.28 USD
76226-494 76226-492
CHIS5-05639
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR