CHIS5-05635

5-05636 5-05635
76226-486EA 393.05 USD
76226-486 76229-836
CHIS5-05635
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR