CHIS5-05633

5-05633 5-05634
76229-832EA 49.13 USD
76229-832 76229-834
CHIS5-05633
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR