CHIS5-05615

5-05615 5-05616
76228-942EA 360.29 USD
76228-942 76228-940
CHIS5-05615
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR