CHIS5-05601

5-05601 5-05602
76229-810EA 589.57 USD
76229-810 76229-808
CHIS5-05601
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR