CHIS5-05569

5-05570 5-05569
76228-882EA 129.38 USD
76228-882 76228-880
CHIS5-05569
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR