CHIS5-05557

5-05557 5-05558
76229-778EA 589.57 USD
76229-778 76229-776
CHIS5-05557
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR