CHIS5-05529

5-05530 5-05529
76228-824EA 114.64 USD
76228-824 76228-826
CHIS5-05529
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR