CHIS5-05503

5-05504 5-05503
76228-774EA 514.24 USD
76228-774 76228-772
CHIS5-05503
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR