CHIS5-05461

5-05462 5-05461
76228-688EA 204.72 USD
76228-688 76228-690
CHIS5-05461
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR