CHIS5-05459

5-05459 5-05460
76228-686EA 499.5 USD
76228-686 76228-684
CHIS5-05459
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR