CHIS5-05457

5-05458 5-05457
76229-752EA 240.75 USD
76229-752 76229-754
CHIS5-05457
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR