CHIS5-05431

5-05432 5-05431
76228-646EA 1310.16 USD
76228-646 76228-644
CHIS5-05431
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR