CHIS5-05385

5-05385 5-05386
76228-554EA 442.18 USD
76228-554 76228-552
CHIS5-05385
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR