CHIS5-05369

5-05370 5-05369
76226-456EA 286.6 USD
76226-456 76226-458
CHIS5-05369
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR