CHIS5-05351

5-05351 5-05352
76228-510EA 917.11 USD
76228-510 76228-508
CHIS5-05351
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR