CHIS5-05345

5-05346 5-05345
76228-498EA 458.56 USD
76228-498 76228-496
CHIS5-05345
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR