CHIS5-05321

5-05322 5-05321
76228-448EA 65.51 USD
76228-448 76228-450
CHIS5-05321
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR