CHIS5-05315

5-05315 5-05316
76226-438EA 491.31 USD
76226-438 76226-436
CHIS5-05315
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR