CHIS5-05275

5-05275 5-05276
76228-390EA 98.26 USD
76228-390 76228-388
CHIS5-05275
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR