CHIS5-05245

5-05245 5-05246
76228-362EA 638.7 USD
76228-362 76228-360
CHIS5-05245
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR